This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00699447_m1
See other TMPRSS12 GE Assays ›
Gene Symbol:
TMPRSS12
Gene Name:
transmembrane protease, serine 12
Gene Aliases:
CT151
Chromosome Location:
Chr.12: 50842918 - 50887884 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00699447_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182559.2 NP_872365.1 2 - 3 435 58
GenBank mRNA BC048112.1 - 2 - 3 420 4830541 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007625.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000398.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMPRSS12

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12004997.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12004997.hg.1 ENST00000551456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12004997.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12004997.hg.1 TC12000398.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12004999.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12004999.hg.1 ENST00000551456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12004999.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12004999.hg.1 TC12000398.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12004998.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12004998.hg.1 ENST00000551456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12004998.hg.1 NM_182559.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12004998.hg.1 TC12000398.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay TC1200007625.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154020.hg.1 TC1200007625.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154022.hg.1 TC1200007625.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154023.hg.1 TC1200007625.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 ENST00000398458.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 ENST00000551456.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 NM_182559.2 Refseq
Affymetrix Human Clariom D Assay PSR1200154021.hg.1 TC1200007625.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0