This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00702141_s1
See other GJB1 GE Assays ›
Gene Symbol:
GJB1
Gene Name:
gap junction protein beta 1
Gene Aliases:
CMTX, CMTX1, CX32
Chromosome Location:
Chr.X: 71215212 - 71225215 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00702141_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000166.5 NP_000157.1 2 568 65
NM_001097642.2 NP_001091111.1 2 517 65
XM_011530907.2 XP_011529209.1 2 616 65
XM_017029408.1 XP_016884897.1 3 737 65
GenBank mRNA AK313474.1 - 2 512 65
BC002805.1 - 2 477 3638623 65
BC022426.1 - 2 548 4252281 65
BC039198.1 - 2 490 4710239 65
BI764356.1 - 2 517 65
BT019329.1 - 1 422 65
X04325.1 - 2 484 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007559.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000384.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Alzheimer's Disease HUMAN SIGNALING IN GAP JUNCTIONS Inventoried Human_Alzheimer Human_Signaling_in_GAP_Junctions Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GJB1

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

transport
cell-cell signaling
nervous system development
purine ribonucleotide transport
gap junction assembly
protein oligomerization
transmembrane transport
gap junction channel activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X005099.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005099.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005099.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X005102.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005102.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005102.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X005105.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005105.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005105.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005105.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005105.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005105.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005094.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005094.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005094.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005094.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X005098.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005098.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005098.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X005106.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005106.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005106.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005106.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005106.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005106.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X005103.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005103.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005103.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00151906.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151906.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151906.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151906.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151906.hg.1 TC0X00007559.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151909.hg.1 TC0X00007559.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151900.hg.1 TC0X00007559.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00151907.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151907.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151907.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151907.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151907.hg.1 TC0X00007559.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151910.hg.1 TC0X00007559.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline