This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00738281_g1
See other TNFRSF25 GE Assays ›
Gene Symbol:
TNFRSF25
Gene Name:
TNF receptor superfamily member 25
Gene Aliases:
APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
Chromosome Location:
Chr.1: 6461151 - 6466195 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00738281_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_148970.1 NP_683871.1 3 - 4 284 95
GenBank mRNA U94508.1 - 3 - 4 196 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100018398.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNFRSF25

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
inflammatory response
immune response
signal transduction
cell surface receptor signaling pathway
multicellular organism development
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
regulation of apoptotic process
positive regulation of MAPK cascade
apoptotic signaling pathway
receptor activity
tumor necrosis factor-activated receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000348333.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000351748.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000351959.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000356876.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000377782.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000414040.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000453260.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000469691.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000473343.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000475730.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000480393.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000485036.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000502588.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000502730.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000510563.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000513135.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_003790.2 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148965.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148966.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148967.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148970.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000348333.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000351748.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000351959.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000356876.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000377782.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000414040.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000453260.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000469691.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000473343.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000475730.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000480393.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000485036.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000502588.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000502730.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000510563.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 ENST00000513135.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_003790.2 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148965.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148966.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148967.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 NM_148970.1 Refseq
Affymetrix Human Clariom D Assay PSR0100241315.hg.1 TC0100018398.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline