This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00739489_m1
See other POLR2K GE Assays ›
Gene Symbol:
POLR2K
Gene Name:
RNA polymerase II subunit K
Gene Aliases:
ABC10-alpha, RPABC4, RPB10alpha, RPB12, RPB7.0, hRPB7.0, hsRPB10a
Chromosome Location:
Chr.8: 100150611 - 100154002 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00739489_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005034.3 NP_005025.1 1 - 2 102 139
GenBank mRNA BC000806.1 - 1 - 2 60 3452723 139
BC018157.1 - 1 - 2 102 3860080 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800008370.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000612.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC000806.1,BC018157.1,NM_005034.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human Transcription of mRNA Inventoried Human_Transcription_of_mRNA Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR2K

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
regulation of transcription from RNA polymerase I promoter
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
transcription from RNA polymerase III promoter
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
positive regulation of type I interferon production
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of gene expression, epigenetic
positive regulation of viral transcription
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08008515.hg.1 TC08000612.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08008515.hg.1 ENST00000353107.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008515.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008515.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008515.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08008514.hg.1 TC08000612.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08008514.hg.1 ENST00000353107.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008514.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008514.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008514.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007571.hg.1 ENST00000553226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006806.hg.1 ENST00000603389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008517.hg.1 TC08000612.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08008517.hg.1 ENST00000353107.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008517.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008517.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08008517.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157637.hg.1 TC0800008370.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157638.hg.1 TC0800008370.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay TC0800008370.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 ENST00000353107.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 ENST00000519765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 ENST00000522439.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 NM_005034.3 Refseq
Affymetrix Human Clariom D Assay PSR0800157641.hg.1 TC0800008370.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200007571.hg.1 ENST00000553226.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006806.hg.1 ENST00000603389.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0