This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00758040_m1
See other ARHGAP6 GE Assays ›
Gene Symbol:
ARHGAP6
Gene Name:
Rho GTPase activating protein 6
Gene Aliases:
RHOGAP6, RHOGAPX-1
Chromosome Location:
Chr.X: 11137543 - 11665701 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00758040_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109776.1 12 - 13 2965 70
GenBank mRNA AF022212.2 - 12 - 13 2966 70
AK303444.1 - 11 - 12 1559 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009069.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
positive regulation of signal transduction
actin filament polymerization
positive regulation of GTPase activity
focal adhesion assembly
regulation of small GTPase mediated signal transduction
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
SH3/SH2 adaptor activity
GTPase activator activity
SH3 domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000303025.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000337414.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000380717.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000380718.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000380736.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000489330.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000495242.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_001287242.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_006125.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_013423.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_013427.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NR_109776.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000303025.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000337414.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000380717.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000380718.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000380736.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000489330.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 ENST00000495242.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_001287242.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_006125.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_013423.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NM_013427.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 NR_109776.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00162194.hg.1 TC0X00009069.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0