This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00765612_g1
See other ALAD GE Assays ›
Gene Symbol:
ALAD
Gene Name:
aminolevulinate dehydratase
Gene Aliases:
ALADH, PBGS
Chromosome Location:
Chr.9: 113386312 - 113401338 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00765612_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALAD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000031.5 NP_000022.3 8 - 9 832 75
NM_001003945.2 NP_001003945.1 8 - 9 998 75
NM_001317745.1 NP_001304674.1 7 - 8 841 75
XM_011518364.2 XP_011516666.1 9 - 10 1024 75
GenBank mRNA AK131490.1 - 8 - 9 971 75
AK290490.1 - 8 - 9 944 75
AK295945.1 - 7 - 8 841 75
AK312552.1 - 8 - 9 780 75
BC000977.3 - 8 - 9 756 2900963 75
M13928.1 - 7 - 8 696 75
S99468.1 - 7 - 8 676 75
S99471.1 - 7 - 8 676 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011260.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001502.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALAD

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to oxidative stress
response to herbicide
response to iron ion
response to zinc ion
response to aluminum ion
response to ionizing radiation
response to vitamin B1
response to selenium ion
response to activity
response to cobalt ion
response to lipopolysaccharide
response to vitamin E
response to drug
response to amino acid
response to ethanol
response to arsenic-containing substance
response to cadmium ion
response to mercury ion
protein homooligomerization
response to glucocorticoid
response to methylmercury
response to platinum ion
response to fatty acid
cellular response to lead ion
cellular response to interleukin-4
catalytic activity
porphobilinogen synthase activity
zinc ion binding
lead ion binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09018745.hg.1 TC09001502.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09018745.hg.1 ENST00000277315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018745.hg.1 ENST00000409155.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018745.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000445750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000445750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000445750.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000445750.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000448137.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000452726.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000464749.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000468504.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482001.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 ENST00000494848.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay TC0900011260.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 ENST00000409155.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 ENST00000482847.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 NM_000031.5 Refseq
Affymetrix Human Clariom D Assay PSR0900176756.hg.1 TC0900011260.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0