This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00766083_m1
See other SACS GE Assays ›
Gene Symbol:
SACS
Gene Name:
sacsin molecular chaperone
Gene Aliases:
ARSACS, DNAJC29, PPP1R138, SPAX6
Chromosome Location:
Chr.13: 23328823 - 23433728 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00766083_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SACS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK090599.1 - 2 - 3 1058 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0