This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00766606_g1
See other PRSS41 GE Assays ›
Gene Symbol:
PRSS41
Gene Name:
protease, serine 41
Gene Aliases:
TESSP1
Chromosome Location:
Chr.16: 2789525 - 2805287 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00766606_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135086.1 NP_001128558.1 2 - 3 296 132
XM_011522477.2 XP_011520779.1 4 - 5 914 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600006635.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000085.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
serine-type peptidase activity
sodium channel regulator activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human CGH Microarray 4x180K 48835 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 122077 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 464978 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 133909 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3416634 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1856 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1857 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5095 - -
Agilent Human Genome CGH + SNP Microarray A_16_P40537099 - -
Agilent Human Genome CGH + SNP Microarray A_16_P40537099 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3416634 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3416634 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30401742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16844239 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891635 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891638 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891639 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891643 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891634 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891637 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891636 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16844227 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891641 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30401756 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891641 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30401756 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891634 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16844239 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16844227 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891638 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891639 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891643 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891636 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891637 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30401742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09891635 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891645 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891676 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891632 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16844225 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891627 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891626 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891647 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891649 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891639 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891634 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891674 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891629 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16844232 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891640 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891652 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891636 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891673 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891641 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891660 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891655 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891671 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891656 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891665 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891638 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891643 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891662 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16844239 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16844228 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09891658 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P40537099 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03103759 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7992717 - -
Affymetrix Human Gene 1.0 ST Array 7992720 - -
Affymetrix Human Gene 1.0 ST Array 7992718 - -
Affymetrix Human Gene 1.0 ST Array 7992719 - -
Affymetrix Human Gene 1.0 ST Array 7992721 - -
Affymetrix Human Gene 1.1 ST Array 7992716 - -
Affymetrix Human Genome U219 Array 11743215_at - -
Affymetrix Human Genome U219 Array 11743215_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR16001594.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001594.hg.1 TC16000085.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16001595.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001595.hg.1 TC16000085.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16001593.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001593.hg.1 TC16000085.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147015.hg.1 TC1600006635.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147017.hg.1 TC1600006635.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147016.hg.1 TC1600006635.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A054210 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A054210 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 203961 - -
Array Name Probe IDs Targets Target Type
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1667596 - -
Illumina human-6 v2.0 expression beadchip ILMN_1667596 - -
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1651230 NM_001135086.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0