This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00766607_m1
See other PRSS41 GE Assays ›
Gene Symbol:
PRSS41
Gene Name:
protease, serine 41
Gene Aliases:
TESSP1
Chromosome Location:
Chr.16: 2789525 - 2805287 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00766607_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135086.1 NP_001128558.1 3 - 4 581 133
XM_011522477.2 XP_011520779.1 5 - 6 1199 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600006635.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000085.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Protease Recommended Assay Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
serine-type peptidase activity
sodium channel regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16001596.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001596.hg.1 TC16000085.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 TC1600006635.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147019.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000399677.2 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 ENST00000421271.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq
Affymetrix Human Clariom D Assay TC1600006635.hg.1 NM_001135086.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline