This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00794896_m1
See other SLC8A3 GE Assays ›
Gene Symbol:
SLC8A3
Gene Name:
solute carrier family 8 member A3
Gene Aliases:
NCX3
Chromosome Location:
Chr.14: 70044217 - 70189626 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00794896_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC8A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104122.1 2 - 3 2531 121
GenBank mRNA AF510503.1 - 2 - 3 2531 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
ion transport
mitochondrial calcium ion transport
cellular calcium ion homeostasis
cell communication
learning
memory
regulation of skeletal muscle contraction
telencephalon development
sodium ion transmembrane transport
myelination
oligodendrocyte differentiation
mitochondrial calcium ion homeostasis
long-term synaptic potentiation
calcium ion transport into cytosol
calcium ion transmembrane transport
cellular response to cAMP
cellular response to hypoxia
regulation of cardiac conduction
calcium ion export from cell
calcium ion import into cell
calcium:sodium antiporter activity
calmodulin binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0