This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00822404_m1
See other ZC3H14 GE Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00822404_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZC3H14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001326295.1 NP_001313224.1 10 - 11 1574 64
NM_001326304.1 NP_001313233.1 9 - 10 1459 64
NM_001326308.1 NP_001313237.1 9 - 10 1459 64
NM_207661.2 NP_997544.1 10 - 11 1422 64
NM_207662.3 NP_997545.2 2 - 3 796 64
XM_005268069.4 XP_005268126.1 10 - 11 1569 64
XM_005268070.4 XP_005268127.1 10 - 11 1569 64
GenBank mRNA AK303123.1 - 11 - 12 1418 64
AL834215.1 - 10 - 11 1422 64
AY578062.1 - 10 - 11 1422 64
BC027607.2 - 2 - 3 782 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400007906.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000536.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZC3H14

Gene Ontology Categories:

Function(s) Process(es)

regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000251038.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000302216.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000318308.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000336693.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000359301.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000380684.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000393514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000393530.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000406216.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000555755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000555900.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000556945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 ENST00000557607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 NM_001160103.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 NM_001160104.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 NM_024824.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 NM_207660.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 NM_207661.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 NM_207662.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006605.hg.1 TC14000536.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000251038.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000302216.12 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000318308.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000336693.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000393514.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000406216.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000554020.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000555755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000555900.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000556000.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000556945.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000557607.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_001160103.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_001160104.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_024824.4 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_207660.3 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_207661.2 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_207662.3 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000251038.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000302216.12 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000318308.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000336693.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000393514.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000406216.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000554020.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000555755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000555900.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000556000.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000556945.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 ENST00000557607.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_001160103.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_001160104.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_024824.4 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_207660.3 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_207661.2 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 NM_207662.3 Refseq
Affymetrix Human Clariom D Assay PSR1400156031.hg.1 TC1400007906.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0