This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00827049_m1
See other ASXL2 GE Assays ›
Gene Symbol:
ASXL2
Gene Name:
additional sex combs like 2, transcriptional regulator
Gene Aliases:
ASXH2
Chromosome Location:
Chr.2: 25733758 - 25878485 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00827049_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASXL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018263.4 NP_060733.4 8 - 9 996 139
XM_006712039.3 XP_006712102.1 4 - 5 3117 139
XM_006712040.2 XP_006712103.1 4 - 5 403 139
XM_011532950.2 XP_011531252.1 7 - 8 1035 139
XM_011532951.2 XP_011531253.1 6 - 7 723 139
XM_017004429.1 XP_016859918.1 8 - 9 826 139
XM_017004430.1 XP_016859919.1 5 - 6 3183 139
GenBank mRNA AB051472.2 - 8 - 9 996 139
AB084279.1 - 8 - 9 859 139
BC042999.2 - 4 - 5 434 5297392 139
BX647527.1 - 9 - 10 1069 139
BX648218.1 - 8 - 9 1021 139
CR933613.1 - 8 - 9 1040 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016645.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001663.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC042999.2,XM_006712040.2,XM_017004430.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ASXL2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of peroxisome proliferator activated receptor signaling pathway
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
DNA binding
peroxisome proliferator activated receptor binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02026601.hg.1 TC02001663.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026601.hg.1 ENST00000272341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026601.hg.1 ENST00000336112.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026601.hg.1 ENST00000404843.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026601.hg.1 ENST00000435504.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026601.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225929.hg.1 TC0200016645.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0