This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00832016_s1
See other ATP5H GE Assays ›
Gene Symbol:
ATP5H
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit D
Gene Aliases:
ATPQ
Chromosome Location:
Chr.17: 75038860 - 75046979 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00832016_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC032245.1 - 1 53 5167128 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC032245.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

ATP synthase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay PSR1700195082.hg.1 TC1700011731.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000580649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 ENST00000582341.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay TC1700011731.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 ENST00000301587.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 ENST00000344546.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 ENST00000538432.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 NM_001003785.1 Refseq
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 NM_006356.2 Refseq
Affymetrix Human Clariom D Assay PSR1700195085.hg.1 TC1700011731.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0