This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00855903_g1
See other PRSS56 GE Assays ›
Gene Symbol:
PRSS56
Gene Name:
protease, serine 56
Gene Aliases:
MCOP6
Chromosome Location:
Chr.2: 232520463 - 232525716 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00855903_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195129.1 NP_001182058.1 5 - 6 681 62
GenBank mRNA BI464671.1 - 2 - 3 235 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200011103.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001401.hg.1

Set Membership:

Amplicon length less than or equal to 70 Protease Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
camera-type eye development
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02022112.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022112.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022110.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022110.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02022113.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022113.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02022108.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022108.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02022111.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022111.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02022109.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022109.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179777.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179780.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179776.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179778.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179775.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179782.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179774.hg.1 TC0200011103.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline