This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00856088_m1
See other PRSS54 GE Assays ›
Gene Symbol:
PRSS54
Gene Name:
protease, serine 54
Gene Aliases:
CT67, KLKBL4
Chromosome Location:
Chr.16: 58279997 - 58295047 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00856088_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS54
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080492.1 NP_001073961.1 5 - 6 919 126
NM_001305173.1 NP_001292102.1 5 - 6 916 126
NM_001305174.1 NP_001292103.1 4 - 5 741 126
GenBank mRNA AK058068.1 - 5 - 6 861 126
AK302175.1 - 4 - 5 671 126
AL137484.1 - 1 - 2 209 126
BC057843.1 - 5 - 6 908 5296090 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600010480.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16001151.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016022.hg.1 ENST00000219301.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016022.hg.1 ENST00000543437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016022.hg.1 NM_001080492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016022.hg.1 TC16001151.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay TC1600010480.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 TC1600010480.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180588.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 NM_001305173.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 TC1600010480.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000219301.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000543437.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000563336.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000567164.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000569079.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 NM_001305174.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 ENST00000569727.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 NM_001080492.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180589.hg.1 NM_001305173.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline