This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00858109_m1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs00858109_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB294925.1 - 1 - 2 Chr. 6 - 110897439 - 110898752 [-] on Build GRCh38 112
GenBankmRNA AK054575.1 - 1 - 2 1673 Chr. 6 - 110896135 - 110898752 [+] on Build GRCh38 112

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adenosylmethionine decarboxylase 1 AMD1 ADOMETDC,
AMD,
SAMDC
-422.0 Sense Chr. 6 - 110814621 - 110895713 on NCBI Build GRCh38
Downstream general transcription factor IIIC subunit 6 GTF3C6 C6orf51,
TFIIIC35,
bA397G5.3
59.8kb Sense Chr. 6 - 110958560 - 110967888 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-859c6f577-lllsl:80/100.66.75.98:80.
git-commit: 13ffd1f44fd3b57634547fe5abf56598aa1cdbac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.0-2026.02.07-1.0