This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00862657_m1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs00862657_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK095401.1 - 1 - 2 43 Chr. 11 - 119612156 - 119618237 [-] on Build GRCh38 91
GenBankmRNA DA445686.1 - 1 - 2 43 Chr. 11 - 119614870 - 119618237 [-] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream USP2 antisense RNA 1 (head to head) USP2-AS1 THY1-AS1 -112.9kb Anti-sense Chr. 11 - 119381778 - 119499233 on NCBI Build GRCh38
Downstream nectin cell adhesion molecule 1 NECTIN1 CD111,
CLPED1,
ED4,
HIgR,
HV1S,
HVEC,
OFC7,
PRR,
PRR1,
PVRL1,
PVRR,
PVRR1,
SK-12,
nectin-1
19.9kb Sense Chr. 11 - 119638098 - 119728725 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0