This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00863083_g1
See other LOC283731 GE Assays ›
Gene Symbol:
LOC283731
Gene Name:
uncharacterized LOC283731
Gene Aliases:
-
Chromosome Location:
Chr.15: 74126373 - 74129278 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00863083_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC283731
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027073.1 1 - 2 315 135
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA719344.1 - 1 - 2 205 Chr. 15 - 74128246 - 74129168 [-] on Build GRCh38 135
GenBankmRNA DA511256.1 - 1 - 2 208 Chr. 15 - 74128285 - 74129171 [-] on Build GRCh38 135
GenBankmRNA DA510716.1 - 1 - 2 208 Chr. 15 - 74128272 - 74129171 [-] on Build GRCh38 135
GenBankmRNA DA790260.1 - 1 - 2 315 Chr. 15 - 74128445 - 74129278 [-] on Build GRCh38 135
GenBankmRNA DA779972.1 - 1 - 2 208 Chr. 15 - 74128267 - 74129171 [-] on Build GRCh38 135
GenBankmRNA DA788519.1 - 1 - 2 179 Chr. 15 - 74128298 - 74129141 [-] on Build GRCh38 136
GenBankmRNA DA713299.1 - 1 - 2 305 Chr. 15 - 74128471 - 74129268 [-] on Build GRCh38 135
GenBankmRNA DA608385.1 - 1 - 2 307 Chr. 15 - 74128363 - 74129270 [-] on Build GRCh38 135
GenBankmRNA DA304005.1 - 1 - 2 210 Chr. 15 - 74128278 - 74129172 [-] on Build GRCh38 135
GenBankmRNA DA614214.1 - 1 - 2 206 Chr. 15 - 74128245 - 74129168 [-] on Build GRCh38 136
GenBankmRNA DA697931.1 - 1 - 2 208 Chr. 15 - 74128160 - 74129171 [-] on Build GRCh38 135
GenBankmRNA DA497913.1 - 1 - 2 214 Chr. 15 - 74128272 - 74129177 [-] on Build GRCh38 135
GenBankmRNA DA788736.1 - 1 - 2 248 Chr. 15 - 74128284 - 74129211 [-] on Build GRCh38 135
GenBankmRNA DA500348.1 - 1 - 2 306 Chr. 15 - 74128357 - 74129268 [-] on Build GRCh38 135
GenBankmRNA DA493344.1 - 1 - 2 183 Chr. 15 - 74128268 - 74129146 [-] on Build GRCh38 135
GenBankmRNA DA483261.1 - 1 - 2 180 Chr. 15 - 74128100 - 74129143 [-] on Build GRCh38 135
GenBankmRNA DA491403.1 - 1 - 2 296 Chr. 15 - 74128342 - 74129259 [-] on Build GRCh38 135
GenBankmRNA DA487880.1 - 1 - 2 277 Chr. 15 - 74128328 - 74129240 [-] on Build GRCh38 135
GenBankmRNA DA777034.1 - 1 - 2 213 Chr. 15 - 74128258 - 74129176 [-] on Build GRCh38 135

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC732265 LOC732265 -27.8kb Sense Chr. 15 - 74088153 - 74100467 on NCBI Build GRCh38
Downstream immunoglobulin superfamily containing leucine rich repeat ISLR HsT17563,
Meflin
44.6kb Anti-sense Chr. 15 - 74173746 - 74176871 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500010003.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001640.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Agilent Affymetrix ArrayStar

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P16805137 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P09758714 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 110994 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 37838 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 407930 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 267201 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 298444 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3718679 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4975 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4974 - -
Agilent Human Genome CGH Microarray 44B A_14_P131807 - -
Agilent Human Genome CGH Microarray 44A A_14_P131807 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15908 - -
Agilent Human Genome CGH + SNP Microarray A_14_P131807 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3718679 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758723 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758763 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758754 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252871 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758753 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758716 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805157 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758726 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805149 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758748 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252862 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758722 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252866 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758756 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758762 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252868 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758744 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758764 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252883 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758765 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758729 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252850 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758728 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758760 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252874 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758724 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252879 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805156 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758733 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252870 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805145 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758758 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758719 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758734 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758752 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758721 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758737 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252858 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252874 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252883 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252862 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758758 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758723 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805156 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252866 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252850 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758753 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758734 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758765 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758722 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758728 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758726 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758721 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758752 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758716 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252868 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252858 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252871 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758733 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758737 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252870 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758744 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805145 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758719 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758748 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252879 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805149 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758762 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758729 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758756 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758760 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758763 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758724 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758764 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30252863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09758754 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16805157 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P131807 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P131807 - -
Agilent Human Genome CGH Microarray 4x44K A_14_P131807 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 75968 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 36168 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P131807 - -
Agilent Human 1B Microarray G4111A A_32_P456880 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16805156 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758745 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758726 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758724 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758716 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758714 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758738 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758750 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758717 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16805147 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758747 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758753 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758740 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758752 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758742 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758733 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16805149 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758762 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758721 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16805143 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758744 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758715 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16805153 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758755 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758727 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758735 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758722 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758730 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758763 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758719 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P09758757 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P131807 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1560723_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR15015921.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15015921.hg.1 NR_027073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15015921.hg.1 TC15001640.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15015922.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15015922.hg.1 NR_027073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15015922.hg.1 TC15001640.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 NR_027073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010003.hg.1 NR_027073.1 Refseq
Affymetrix Human Clariom D Assay PSR1500174477.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500174477.hg.1 NR_027073.1 Refseq
Affymetrix Human Clariom D Assay PSR1500174477.hg.1 TC1500010003.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500174477.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500174477.hg.1 NR_027073.1 Refseq
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 NR_027073.1 Refseq
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 TC1500010003.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500174475.hg.1 NR_027073.1 Refseq
Affymetrix Human Clariom D Assay TC1500010003.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010003.hg.1 ENST00000514871.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010003.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010003.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010003.hg.1 ENST00000563727.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010003.hg.1 NR_027073.1 Refseq
Affymetrix Human Clariom D Assay TC1500010003.hg.1 NR_027073.1 Refseq
Human Genome U133 Plus 2.0 Array 1560723_PM_at - -
Human Genome U133 Plus 2.0 Array 1560723_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560723_at11 - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A052372 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A052372 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline