This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00872105_g1
See other POLRMT GE Assays ›
Gene Symbol:
POLRMT
Gene Name:
RNA polymerase mitochondrial
Gene Aliases:
APOLMT, MTRNAP, MTRPOL, h-mtRPOL
Chromosome Location:
Chr.19: 617221 - 633592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00872105_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLRMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005035.3 NP_005026.3 9 - 10 1911 64
XM_005259580.3 XP_005259637.2 9 - 10 1935 64
GenBank mRNA AB209920.1 - 7 - 8 1383 64
AK302877.1 - 9 - 10 1910 64
AK307525.1 - 9 - 10 1856 64
BC030829.1 - 9 - 10 1863 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900009100.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000987.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLRMT

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from mitochondrial promoter
transcription initiation from mitochondrial promoter
mitochondrion organization
DNA binding
DNA-directed RNA polymerase activity
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19013454.hg.1 ENST00000215591.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19013454.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19013454.hg.1 TC19000987.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19013455.hg.1 ENST00000215591.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19013455.hg.1 NM_005035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19013455.hg.1 TC19000987.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000587057.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000589961.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590336.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590573.4 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000590709.3 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592633.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 ENST00000592863.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay TC1900009100.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay PSR1900172242.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900172242.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay PSR1900172242.hg.1 TC1900009100.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900172242.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900172242.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay PSR1900172241.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900172241.hg.1 NM_005035.3 Refseq
Affymetrix Human Clariom D Assay PSR1900172241.hg.1 TC1900009100.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900172241.hg.1 ENST00000588649.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900172241.hg.1 NM_005035.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0