This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00872623_m1
See other FREM2 GE Assays ›
Gene Symbol:
FREM2
Gene Name:
FRAS1 related extracellular matrix protein 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 38687036 - 38887131 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00872623_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FREM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207361.5 NP_997244.4 3 - 4 5717 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300006919.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000132.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FREM2

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
cell communication
cell adhesion
heart development
inner ear development
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13001523.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001523.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001523.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13001528.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001528.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001528.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001528.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13001529.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001529.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001529.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13001522.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001522.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001522.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13001524.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001524.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001524.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13001526.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001526.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001526.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001526.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13001527.hg.1 ENST00000280481.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001527.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13001527.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13001527.hg.1 TC13000132.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300147008.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147008.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147008.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147008.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147008.hg.1 TC1300006919.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300147006.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147006.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147006.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147006.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147006.hg.1 TC1300006919.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147012.hg.1 TC1300006919.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300147013.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147013.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147013.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147013.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147013.hg.1 TC1300006919.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147011.hg.1 TC1300006919.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300147007.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147007.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147007.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147007.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147007.hg.1 TC1300006919.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay PSR1300147010.hg.1 TC1300006919.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0