This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00877853_g1
See other LOC107983954 GE Assays ›
Gene Symbol:
LOC107983954
Gene Name:
extracellular matrix protein 2-like
Gene Aliases:
-
Chromosome Location:
Chr.X: 153476926 - 153483800 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00877853_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC107983954
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017030018.1 XP_016885507.1 3 - 4 4972 84
XM_017030019.1 XP_016885508.1 4 - 5 4773 84
XM_017030020.1 XP_016885509.1 4 - 5 4617 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-89699:80/100.66.75.179:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0