This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00893433_g1
See other MFSD2A GE Assays ›
Gene Symbol:
MFSD2A
Gene Name:
major facilitator superfamily domain containing 2A
Gene Aliases:
MCPH15, MFSD2, NLS1
Chromosome Location:
Chr.1: 39955112 - 39969968 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00893433_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MFSD2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136493.2 NP_001129965.1 9 - 10 1231 74
NM_001287808.1 NP_001274737.1 7 - 8 932 74
NM_001287809.1 NP_001274738.1 9 - 10 1075 74
NM_032793.4 NP_116182.2 9 - 10 1192 74
XM_005271285.1 XP_005271342.1 9 - 10 1156 74
GenBank mRNA AB073383.1 - 9 - 10 1156 74
AF289609.1 - 9 - 10 1148 74
AK074186.1 - 9 - 10 1097 74
AK075183.1 - 9 - 10 1159 74
AK093223.1 - 9 - 10 1193 74
AK291540.1 - 9 - 10 1154 74
AK297991.1 - 9 - 10 1039 74
AK301533.1 - 7 - 8 899 74
AY358636.1 - 9 - 10 1174 74
BC006353.2 - 5 - 6 527 74
BC011587.1 - 9 - 10 1166 4646430 74
BC092414.1 - 9 - 10 1134 30387073 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007936.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000503.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MFSD2A

Gene Ontology Categories:

Function(s) Process(es)

fatty acid transport
phospholipid transport
transcytosis
transmembrane transport
establishment of blood-brain barrier
lipid transport across blood brain barrier
phospholipid transporter activity
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 TC01000503.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 ENST00000372809.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 ENST00000372811.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 ENST00000420632.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 ENST00000480630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 NM_001136493.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007803.hg.1 NM_032793.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000372809.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000372811.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000420632.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000459917.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000480630.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000481612.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000491515.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_001136493.2 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_001287808.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_001287809.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_032793.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000372809.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000372811.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000420632.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000459917.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000480630.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000481612.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 ENST00000491515.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_001136493.2 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_001287808.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_001287809.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 NM_032793.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155820.hg.1 TC0100007936.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0