This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00894474_m1
See other DNASE1L3 GE Assays ›
Gene Symbol:
DNASE1L3
Gene Name:
deoxyribonuclease I like 3
Gene Aliases:
DHP2, DNAS1L3, LSD, SLEB16
Chromosome Location:
Chr.3: 58192626 - 58211003 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00894474_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNASE1L3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256560.1 NP_001243489.1 5 - 6 709 85
NM_004944.3 NP_004935.1 6 - 7 799 85
GenBank mRNA AB209613.1 - 3 - 4 3325 85
AF047354.1 - 6 - 7 772 85
AI591302.1 - 2 - 3 85
AI660448.1 - 2 - 3 85
AK131323.1 - 9 - 10 1294 85
AK301263.1 - 5 - 6 675 85
AK313303.1 - 6 - 7 765 85
BC015831.1 - 6 - 7 769 4723725 85
DB164811.1 - 3 - 4 296 85
U56814.1 - 6 - 7 726 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300011298.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001498.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DNASE1L3

Gene Ontology Categories:

Function(s) Process(es)

DNA metabolic process
apoptotic DNA fragmentation
programmed cell death involved in cell development
DNA binding
endodeoxyribonuclease activity
deoxyribonuclease activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 ENST00000318316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 ENST00000394549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 ENST00000450710.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 ENST00000483681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 ENST00000486455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03027295.hg.1 TC03001498.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000463694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000463694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000463694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03027297.hg.1 ENST00000318316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027297.hg.1 ENST00000394549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027297.hg.1 ENST00000450710.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027297.hg.1 ENST00000483681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03027297.hg.1 NM_004944.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03027297.hg.1 TC03001498.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay PSR0300184741.hg.1 TC0300011298.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000394549.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000460422.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000461914.7 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000463694.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000463694.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000463694.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000477209.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000483681.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 ENST00000486455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_001256560.1 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq
Affymetrix Human Clariom D Assay TC0300011298.hg.1 NM_004944.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline