This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00894502_g1
See other PLCH1 GE Assays ›
Gene Symbol:
PLCH1
Gene Name:
phospholipase C eta 1
Gene Aliases:
PLCL3
Chromosome Location:
Chr.3: 155458603 - 155745723 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00894502_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLCH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130961.1 NP_001124433.1 23 - 24 3301 164
XM_011512567.1 XP_011510869.1 24 - 25 3350 164
GenBank mRNA AB028992.1 - 19 - 20 2379 164
AY691170.1 - 23 - 24 3024 164

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
inositol phosphate metabolic process
phosphatidylinositol-mediated signaling
phosphatidylinositol phospholipase C activity
signal transducer activity
calcium ion binding
calcium-dependent phospholipase C activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0