This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00895485_g1
See other LPGAT1 GE Assays ›
Gene Symbol:
LPGAT1
Gene Name:
lysophosphatidylglycerol acyltransferase 1
Gene Aliases:
FAM34A, FAM34A1, NET8
Chromosome Location:
Chr.1: 211743457 - 211830772 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00895485_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LPGAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320808.1 NP_001307737.1 3 - 4 1178 143
NM_014873.2 NP_055688.1 3 - 4 589 143
XM_005273362.3 XP_005273419.1 4 - 5 737 143
XM_011510228.2 XP_011508530.1 4 - 5 626 143
XM_011510229.2 XP_011508531.1 3 - 4 509 143
XM_017003004.1 XP_016858493.1 2 - 3 1741 143
GenBank mRNA AY561706.1 - 2 - 3 362 143
BC034621.1 - 3 - 4 1179 4839400 143
D86960.1 - 3 - 4 589 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100017223.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003800.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LPGAT1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
phospholipid biosynthetic process
phosphatidylglycerol acyl-chain remodeling
molecular_function
1-acylglycerol-3-phosphate O-acyltransferase activity
2-acylglycerol-3-phosphate O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01058108.hg.1 TC01003800.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01058108.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01058108.hg.1 ENST00000366997.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01058108.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000498690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000498690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 ENST00000498690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay PSR0100230417.hg.1 TC0100017223.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366996.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000366997.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000488600.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000498690.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000498690.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 ENST00000498690.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq
Affymetrix Human Clariom D Assay TC0100017223.hg.1 NM_014873.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0