This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00895616_m1
See other FH GE Assays ›
Gene Symbol:
FH
Gene Name:
fumarate hydratase
Gene Aliases:
FMRD, HLRCC, LRCC, MCL, MCUL1
Chromosome Location:
Chr.1: 241497557 - 241519785 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00895616_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000143.3 NP_000134.2 2 - 3 329 93
XM_011544132.2 XP_011542434.1 2 - 3 769 93
GenBank mRNA AK312415.1 - 2 - 3 305 93
BC003108.1 - 2 - 3 291 3502012 93
BC017444.1 - 2 - 3 281 3606118 93
BT009839.1 - 2 - 3 266 93
DA129148.1 - 2 - 3 329 93
KU178071.1 - 2 - 3 266 93
KU178072.1 - 2 - 3 266 93
KU178073.1 - 2 - 3 266 93
KU178074.1 - 2 - 3 266 93
KU178075.1 - 2 - 3 266 93
U48857.1 - 2 - 3 296 93
U59309.1 - 2 - 3 299 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100017945.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01004025.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_011544132.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FH

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
fumarate metabolic process
malate metabolic process
homeostasis of number of cells within a tissue
protein tetramerization
fumarate hydratase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01060805.hg.1 TC01004025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01060805.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01060805.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01060805.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01060803.hg.1 TC01004025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01060803.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01060803.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235154.hg.1 TC0100017945.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235152.hg.1 TC0100017945.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0