This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00895617_m1
See other FH GE Assays ›
Gene Symbol:
FH
Gene Name:
fumarate hydratase
Gene Aliases:
FMRD, HLRCC, LRCC, MCL, MCUL1
Chromosome Location:
Chr.1: 241497557 - 241519785 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00895617_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000143.3 NP_000134.2 3 - 4 440 160
XM_011544132.2 XP_011542434.1 3 - 4 880 160
GenBank mRNA AK312415.1 - 3 - 4 416 160
BC003108.1 - 3 - 4 402 3502012 160
BC017444.1 - 3 - 4 392 3606118 160
BT009839.1 - 3 - 4 377 160
KU178071.1 - 3 - 4 377 160
KU178073.1 - 3 - 4 377 160
M15502.1 - 2 - 3 248 160
U48857.1 - 3 - 4 407 160
U59309.1 - 3 - 4 410 160

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100017945.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01004025.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FH

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
fumarate metabolic process
malate metabolic process
homeostasis of number of cells within a tissue
protein tetramerization
fumarate hydratase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01060802.hg.1 TC01004025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01060802.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01060802.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay PSR0100235151.hg.1 TC0100017945.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000366560.3 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000493477.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 ENST00000497042.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq
Affymetrix Human Clariom D Assay TC0100017945.hg.1 NM_000143.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0