This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00895796_g1
See other ABCG8 GE Assays ›
Gene Symbol:
ABCG8
Gene Name:
ATP binding cassette subfamily G member 8
Gene Aliases:
GBD4, STSL
Chromosome Location:
Chr.2: 43831942 - 43880051 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00895796_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022437.2 NP_071882.1 9 - 10 1503 73
XM_005264483.3 XP_005264540.1 9 - 10 1615 73
XM_011533029.2 XP_011531331.1 9 - 10 2070 73
XM_011533030.2 XP_011531332.1 9 - 10 1912 73
GenBank mRNA AF320294.1 - 9 - 10 1413 73
AF324494.1 - 9 - 10 1503 73
BC113657.1 - 9 - 10 1449 8322709 73
BC143564.1 - 9 - 10 1446 9052075 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200007402.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000272.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABCG8

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

response to nutrient
excretion
negative regulation of intestinal phytosterol absorption
phospholipid transport
sterol transport
intestinal cholesterol absorption
cholesterol efflux
response to drug
cholesterol homeostasis
negative regulation of intestinal cholesterol absorption
transmembrane transport
protein binding
ATP binding
ATPase activity
cholesterol transporter activity
ATPase activity, coupled to transmembrane movement of substances
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02004064.hg.1 TC02000272.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02004064.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02004064.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay TC0200007402.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay PSR0200151281.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200151281.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay PSR0200151281.hg.1 ENST00000272286.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200151281.hg.1 NM_022437.2 Refseq
Affymetrix Human Clariom D Assay PSR0200151281.hg.1 TC0200007402.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0