This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00899695_m1
See other CDH4 GE Assays ›
Gene Symbol:
CDH4
Gene Name:
cadherin 4
Gene Aliases:
CAD4, R-CAD, RCAD
Chromosome Location:
Chr.20: 61252426 - 61940617 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00899695_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252338.2 NP_001239267.1 11 - 12 1903 62
NM_001252339.2 NP_001239268.1 11 - 12 2060 62
NM_001794.4 NP_001785.2 12 - 13 2088 62
GenBank mRNA AK091496.1 - 12 - 13 2284 62
AK124724.1 - 11 - 12 2043 62
AK308345.1 - 11 - 12 1902 62
AK315192.1 - 12 - 13 2042 62
BC101651.1 - 12 - 13 2088 8069157 62
BC112150.1 - 12 - 13 2088 8327618 62
L34059.1 - 12 - 13 2014 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007973.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000481.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH4

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
axon guidance
adherens junction organization
positive regulation of axon extension
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 ENST00000360469.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 ENST00000536643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 ENST00000543233.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 NM_001252338.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 NM_001794.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20006703.hg.1 TC20000481.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay TC2000007973.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 NM_001252339.2 Refseq
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 TC2000007973.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 ENST00000543233.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 ENST00000611855.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 ENST00000614565.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000155486.hg.1 NM_001252339.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-2vf2v:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline