This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00899863_m1
See other GFPT1 GE Assays ›
Gene Symbol:
GFPT1
Gene Name:
glutamine--fructose-6-phosphate transaminase 1
Gene Aliases:
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG
Chromosome Location:
Chr.2: 69319769 - 69387254 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00899863_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GFPT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244710.1 NP_001231639.1 17 - 18 1913 103
NM_002056.3 NP_002047.2 16 - 17 1859 103
XM_017003801.1 XP_016859290.1 17 - 18 1877 103
XM_017003802.1 XP_016859291.1 16 - 17 1815 103
GenBank mRNA BC045641.1 - 16 - 17 1855 5298728 103
M90516.1 - 16 - 17 1798 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012956.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001948.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GFPT1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
fructose 6-phosphate metabolic process
glucosamine biosynthetic process
UDP-N-acetylglucosamine metabolic process
UDP-N-acetylglucosamine biosynthetic process
energy reserve metabolic process
glutamine metabolic process
response to sucrose
cellular response to insulin stimulus
circadian regulation of gene expression
IRE1-mediated unfolded protein response
negative regulation of glycogen biosynthetic process
protein homotetramerization
glutamine-fructose-6-phosphate transaminase (isomerizing) activity
amino acid binding
carbohydrate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02030871.hg.1 TC02001948.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02030871.hg.1 ENST00000357308.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02030871.hg.1 ENST00000361060.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02030871.hg.1 NM_001244710.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02030871.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000493759.2 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 ENST00000494201.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay TC0200012956.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 ENST00000357308.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 ENST00000361060.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 NM_001244710.1 Refseq
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 NM_002056.3 Refseq
Affymetrix Human Clariom D Assay PSR0200194132.hg.1 TC0200012956.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0