This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00900359_m1
See other TNFRSF11B GE Assays ›
Gene Symbol:
TNFRSF11B
Gene Name:
TNF receptor superfamily member 11b
Gene Aliases:
OCIF, OPG, PDB5, TR1
Chromosome Location:
Chr.8: 118923557 - 118952144 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00900359_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF11B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002546.3 NP_002537.3 3 - 4 920 107
GenBank mRNA AB002146.1 - 3 - 4 597 107
AF134187.1 - 2 - 3 535 107
AK223155.1 - 3 - 4 659 107
AK308524.1 - 4 - 5 941 107
AK313710.1 - 3 - 4 1014 107
BC030155.2 - 3 - 4 818 4875170 107
U94332.1 - 3 - 4 691 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011611.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001552.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNFRSF11B

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
inflammatory response
immune response
signal transduction
response to nutrient
extracellular matrix organization
response to magnesium ion
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
negative regulation of odontogenesis of dentin-containing tooth
response to drug
regulation of apoptotic process
positive regulation of MAPK cascade
response to estrogen
negative regulation of bone resorption
response to arsenic-containing substance
apoptotic signaling pathway
receptor activity
tumor necrosis factor-activated receptor activity
cytokine activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08020440.hg.1 TC08001552.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08020440.hg.1 ENST00000297350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08020440.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay PSR0800178791.hg.1 TC0800011611.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000297350.8 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000517352.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 ENST00000521597.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq
Affymetrix Human Clariom D Assay TC0800011611.hg.1 NM_002546.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0