This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00900721_m1
See other SGPL1 GE Assays ›
Gene Symbol:
SGPL1
Gene Name:
sphingosine-1-phosphate lyase 1
Gene Aliases:
S1PL, SPL
Chromosome Location:
Chr.10: 70815920 - 70881189 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00900721_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SGPL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003901.3 NP_003892.2 12 - 13 1525 92
XM_005270263.1 XP_005270320.1 12 - 13 1538 92
XM_006718053.1 XP_006718116.1 13 - 14 1640 92
XM_011540316.2 XP_011538618.1 12 - 13 1584 92
XM_011540317.1 XP_011538619.1 12 - 13 1434 92
XM_011540318.1 XP_011538620.1 11 - 12 1455 92
GenBank mRNA AB033078.1 - 12 - 13 1503 92
AF144638.1 - 11 - 12 1303 92
AJ011304.2 - 12 - 13 1480 92
AK314615.1 - 12 - 13 1434 92
BC052991.1 - 12 - 13 1513 6150776 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000007936.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000434.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SGPL1

Gene Ontology Categories:

Function(s) Process(es)

luteinization
vasculogenesis
kidney development
fatty acid metabolic process
ceramide metabolic process
spermatogenesis
androgen metabolic process
estrogen metabolic process
post-embryonic development
fibroblast migration
hemopoiesis
sphingolipid biosynthetic process
sphingolipid catabolic process
Leydig cell differentiation
regulation of multicellular organism growth
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
palate development
face morphogenesis
apoptotic signaling pathway
protein binding
sphinganine-1-phosphate aldolase activity
carboxy-lyase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10005104.hg.1 ENST00000373202.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005104.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005104.hg.1 TC10000434.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10005103.hg.1 ENST00000373202.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005103.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005103.hg.1 TC10000434.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153886.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153886.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153886.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153886.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153886.hg.1 TC1000007936.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0