Assay Details
| Gene Symbol: CDH26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528969.2 | XP_011527271.1 | 17 - 18 | 2619 | 97 | |
| XM_011528970.2 | XP_011527272.1 | 11 - 12 | 1635 | 97 | ||
| XM_017027994.1 | XP_016883483.1 | 17 - 18 | 3282 | 97 | ||
| XM_017027995.1 | XP_016883484.1 | 16 - 17 | 3214 | 97 | ||
| XM_017027996.1 | XP_016883485.1 | 17 - 18 | 3184 | 97 | ||
| XM_017027997.1 | XP_016883486.1 | 15 - 16 | 3160 | 97 | ||
| XM_017028000.1 | XP_016883489.1 | 16 - 17 | 2452 | 97 | ||
| GenBank mRNA | AF169690.1 | - | 7 - 8 | 954 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |

CDH26
Genomic Map