This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902331_m1
See other CDH26 GE Assays ›
Gene Symbol:
CDH26
Gene Name:
cadherin 26
Gene Aliases:
VR20
Chromosome Location:
Chr.20: 59957764 - 60035629 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00902331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177980.2 NP_817089.1 10 - 11 1732 98
XM_011528969.2 XP_011527271.1 10 - 11 1738 98
XM_011528970.2 XP_011527272.1 4 - 5 754 98
XM_017027994.1 XP_016883483.1 10 - 11 2401 98
XM_017027995.1 XP_016883484.1 10 - 11 2402 98
XM_017027996.1 XP_016883485.1 10 - 11 2303 98
XM_017027997.1 XP_016883486.1 10 - 11 2402 98
XM_017027998.1 XP_016883487.1 10 - 11 2399 98
XM_017027999.1 XP_016883488.1 10 - 11 2399 98
XM_017028000.1 XP_016883489.1 9 - 10 1571 98
XM_017028001.1 XP_016883490.1 10 - 11 2399 98
XM_017028002.1 XP_016883491.1 10 - 11 2400 98
GenBank mRNA AF169690.1 - 2 - 3 196 98
AK055202.1 - 10 - 11 1721 98
BC136306.1 - 10 - 11 1536 9020293 98
BC143931.1 - 10 - 11 1536 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007943.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000474.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH26

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20006606.hg.1 ENST00000244047.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20006606.hg.1 ENST00000348616.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20006606.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20006606.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20006606.hg.1 TC20000474.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 TC2000007943.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000155266.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000244049.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000348616.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000350849.10 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000370991.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000456106.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000477058.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 ENST00000497614.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_021810.5 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq
Affymetrix Human Clariom D Assay TC2000007943.hg.1 NM_177980.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zsmkz:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0