This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902572_g1
See other TNFSF13B GE Assays ›
Gene Symbol:
TNFSF13B
Gene Name:
tumor necrosis factor superfamily member 13b
Gene Aliases:
BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, TNLG7A, ZTNF4
Chromosome Location:
Chr.13: 108268240 - 108308484 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00902572_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFSF13B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145645.2 NP_001139117.1 1 - 2 607 77
NM_006573.4 NP_006564.1 1 - 2 607 77
GenBank mRNA AF116456.1 - 1 - 2 458 77
AF132600.1 - 1 - 2 476 77
AF134715.1 - 1 - 2 484 77
AF136293.1 - 1 - 2 340 77
AF186114.1 - 1 - 2 497 77
AK309629.1 - 2 - 3 377 77
AY129225.1 - 1 - 2 340 77
AY129226.1 - 1 - 2 109 77
AY302751.1 - 1 - 2 340 77
AY358881.1 - 1 - 2 509 77
BC020674.1 - 1 - 2 519 4719315 77
BI818459.1 - 1 - 2 607 77
CR541818.1 - 1 - 2 340 77
HM636064.1 - 1 - 2 340 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNFSF13B

Gene Ontology Categories:

Function(s) Process(es)

immune response
signal transduction
cell proliferation
positive regulation of cell proliferation
positive regulation of B cell proliferation
tumor necrosis factor-mediated signaling pathway
receptor binding
cytokine activity
tumor necrosis factor receptor binding
protein binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 177543 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline