This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902575_g1
See other TNFSF13B GE Assays ›
Gene Symbol:
TNFSF13B
Gene Name:
tumor necrosis factor superfamily member 13b
Gene Aliases:
BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, TNLG7A, ZTNF4
Chromosome Location:
Chr.13: 108268240 - 108308484 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00902575_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFSF13B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145645.2 NP_001139117.1 3 - 4 792 128
NM_006573.4 NP_006564.1 4 - 5 849 128
GenBank mRNA AF116456.1 - 4 - 5 700 128
AF132600.1 - 4 - 5 718 128
AF134715.1 - 4 - 5 726 128
AF136293.1 - 4 - 5 582 128
AF186114.1 - 4 - 5 739 128
AK309629.1 - 5 - 6 128
AY129225.1 - 4 - 5 582 128
AY129226.1 - 4 - 5 351 128
AY129227.1 - 3 - 4 249 128
AY129228.1 - 3 - 4 201 128
AY302751.1 - 3 - 4 525 128
AY358881.1 - 4 - 5 751 128
BC020674.1 - 4 - 5 761 4719315 128
CR541818.1 - 4 - 5 582 128

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNFSF13B

Gene Ontology Categories:

Function(s) Process(es)

immune response
signal transduction
cell proliferation
positive regulation of cell proliferation
positive regulation of B cell proliferation
tumor necrosis factor-mediated signaling pathway
receptor binding
cytokine activity
tumor necrosis factor receptor binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline