This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902792_m1
See other ALDOC GE Assays ›
Gene Symbol:
ALDOC
Gene Name:
aldolase, fructose-bisphosphate C
Gene Aliases:
ALDC
Chromosome Location:
Chr.17: 28573115 - 28576963 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00902792_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005257949.2 XP_005258006.1 1 - 2 163 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005257949.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

fructose metabolic process
gluconeogenesis
fructose 1,6-bisphosphate metabolic process
epithelial cell differentiation
canonical glycolysis
fructose-bisphosphate aldolase activity
protein binding
cytoskeletal protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000226253.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395319.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000395321.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000435638.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000460201.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000578590.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000581807.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000582381.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 ENST00000584086.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq
Affymetrix Human Clariom D Assay TC1700010200.hg.1 NM_005165.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0