| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
ENST00000518689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
ENST00000520948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
ENST00000523873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001025366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001025367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001171623.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001171624.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001171625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001204384.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_001204385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
NM_003376.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007307.hg.1 |
TC06000608.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000372055.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000413642.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000425836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000480614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000518689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000519767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000520948.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000523873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000611736.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001025366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001025367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001171623.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001171624.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001171625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001204384.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001204385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_003376.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000372055.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000413642.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000425836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000480614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000518689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000519767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000520948.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000523873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
ENST00000611736.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001025366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001025367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001171623.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001171624.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001171625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001204384.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_001204385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
NM_003376.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156672.hg.1 |
TC0600008109.hg.1 |
Clariom D TC |