| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019364.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019364.hg.1 |
ENST00000451316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019364.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019364.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019364.hg.1 |
TC06001592.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011561.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
ENST00000293756.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
ENST00000451316.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
NM_001142883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
NM_054111.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179229.hg.1 |
TC0600011561.hg.1 |
Clariom D TC |