This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00903563_g1
See other LDHD GE Assays ›
Gene Symbol:
LDHD
Gene Name:
lactate dehydrogenase D
Gene Aliases:
DLD
Chromosome Location:
Chr.16: 75111860 - 75116772 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00903563_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LDHD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153486.3 NP_705690.2 5 - 6 680 116
GenBank mRNA BC040279.1 - 5 - 6 645 5162826 116
BC047902.1 - 5 - 6 675 5762691 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600010866.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16001274.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LDHD

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
D-lactate dehydrogenase (cytochrome) activity
protein binding
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16017752.hg.1 ENST00000300051.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16017752.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16017752.hg.1 TC16001274.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay PSR1600184514.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600184514.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay PSR1600184514.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600184514.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay PSR1600184514.hg.1 TC1600010866.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000300051.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000450168.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000568164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 ENST00000569876.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_153486.3 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq
Affymetrix Human Clariom D Assay TC1600010866.hg.1 NM_194436.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline