This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00905219_m1
See other NPC1L1 GE Assays ›
Gene Symbol:
NPC1L1
Gene Name:
NPC1 like intracellular cholesterol transporter 1
Gene Aliases:
NPC11L1
Chromosome Location:
Chr.7: 44512440 - 44541650 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00905219_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPC1L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101648.1 NP_001095118.1 10 - 11 2693 110
NM_013389.2 NP_037521.2 10 - 11 2693 110
XM_011515326.2 XP_011513628.1 9 - 10 2544 110
XM_011515327.2 XP_011513629.1 10 - 11 2948 110
XM_011515328.2 XP_011513630.1 7 - 8 2292 110
GenBank mRNA AF192522.1 - 10 - 11 2693 110
AY437865.1 - 10 - 11 2637 110
AY515256.1 - 10 - 11 2637 110
BC117178.1 - 10 - 11 2691 40125729 110
FJ481111.1 - 10 - 11 2637 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010924.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001342.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NPC1L1

Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
sterol transport
intestinal cholesterol absorption
cholesterol transport
lipoprotein metabolic process
response to drug
protein binding
drug binding
Rab GTPase binding
myosin V binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000289547.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000381160.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000423141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 ENST00000546276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001101648.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_001300967.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010924.hg.1 NM_013389.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0