This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00905771_m1
See other SMURF1 GE Assays ›
Gene Symbol:
SMURF1
Gene Name:
SMAD specific E3 ubiquitin protein ligase 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 99027435 - 99144120 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00905771_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMURF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020429.2 NP_065162.1 9 - 10 1222 91
XM_017012457.1 XP_016867946.1 9 - 10 1220 91
GenBank mRNA AB046845.1 - 9 - 10 1190 91
AF464850.1 - 9 - 10 882 91
BC152468.1 - 9 - 10 1190 8862605 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700011929.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001639.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMURF1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
positive regulation of defense response to virus by host
protein export from nucleus
transforming growth factor beta receptor signaling pathway
ectoderm development
protein ubiquitination
cell differentiation
negative regulation of ossification
BMP signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
ubiquitin-dependent SMAD protein catabolic process
receptor catabolic process
protein localization to cell surface
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
Wnt signaling pathway, planar cell polarity pathway
engulfment of target by autophagosome
protein targeting to vacuole involved in autophagy
protein targeting to plasma membrane
mitophagy in response to mitochondrial depolarization
positive regulation of dendrite extension
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
phospholipid binding
ligase activity
activin binding
ubiquitin protein ligase activity
I-SMAD binding
R-SMAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07024999.hg.1 TC07001639.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07024999.hg.1 ENST00000361125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024999.hg.1 ENST00000361368.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024999.hg.1 NM_001199847.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07024999.hg.1 NM_020429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07024999.hg.1 NM_181349.2 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 ENST00000361125.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 ENST00000361368.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 NM_001199847.1 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 NM_020429.2 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 NM_181349.2 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 ENST00000361125.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 ENST00000361368.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 NM_001199847.1 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 NM_020429.2 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 NM_181349.2 Refseq
Affymetrix Human Clariom D Assay PSR0700185303.hg.1 TC0700011929.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0