This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00905808_m1
See other PLA2G12B GE Assays ›
Gene Symbol:
PLA2G12B
Gene Name:
phospholipase A2 group XIIB
Gene Aliases:
FKSG71, GXIIB, GXIIIsPLA2, PLA2G13, sPLA2-GXIIB
Chromosome Location:
Chr.10: 72934762 - 72955044 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00905808_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA2G12B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318124.1 NP_001305053.1 2 - 3 437 66
NM_032562.4 NP_115951.2 2 - 3 437 66
GenBank mRNA AF339053.1 - 2 - 3 396 66
AF349540.1 - 2 - 3 340 66
AK313712.1 - 2 - 3 400 66
AY358032.1 - 2 - 3 370 66
BC093996.1 - 2 - 3 359 7939841 66
BC111946.1 - 2 - 3 359 8327414 66
BC143532.1 - 2 - 3 359 9052041 66
BP274516.1 - 2 - 3 437 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011003.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLA2G12B

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
phospholipase A2 activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay TC1000011003.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay PSR1000174837.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000174837.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay PSR1000174837.hg.1 ENST00000373032.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000174837.hg.1 NM_032562.4 Refseq
Affymetrix Human Clariom D Assay PSR1000174837.hg.1 TC1000011003.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fxpwm:80/100.66.73.1:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0