This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00905824_gH
See other GOT2 GE Assays ›
Gene Symbol:
GOT2
Gene Name:
glutamic-oxaloacetic transaminase 2
Gene Aliases:
KAT4, KATIV, KYAT4, mitAAT
Chromosome Location:
Chr.16: 58707131 - 58734357 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00905824_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286220.1 NP_001273149.1 3 - 4 435 75
NM_002080.3 NP_002071.2 4 - 5 564 75
GenBank mRNA AK098313.1 - 4 - 5 486 75
AK223271.1 - 4 - 5 499 75
AK295993.1 - 3 - 4 404 75
AK307598.1 - 4 - 5 451 75
AL516773.3 - 4 - 5 549 75
BC000525.2 - 4 - 5 487 2961181 75
BI667777.1 - 4 - 5 569 75
M22632.1 - 4 - 5 443 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600010500.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16001158.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GOT2

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
2-oxoglutarate metabolic process
oxaloacetate metabolic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
glutamate metabolic process
cellular amino acid biosynthetic process
fatty acid transport
4-hydroxyproline catabolic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
response to ethanol
L-kynurenine metabolic process
L-aspartate:2-oxoglutarate aminotransferase activity
phospholipid binding
kynurenine-oxoglutarate transaminase activity
amino acid binding
enzyme binding
pyridoxal phosphate binding
identical protein binding
protein homodimerization activity
poly(A) RNA binding
L-phenylalanine:2-oxoglutarate aminotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016132.hg.1 ENST00000245206.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016132.hg.1 ENST00000425685.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016132.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016132.hg.1 NM_002080.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016132.hg.1 TC16001158.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000492378.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000494627.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay TC1600010500.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 NM_002080.3 Refseq
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 TC1600010500.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000245206.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000434819.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000496461.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000564400.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 ENST00000568368.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 NM_001286220.1 Refseq
Affymetrix Human Clariom D Assay PSR1600180807.hg.1 NM_002080.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline