This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00906891_m1
See other CDH20 GE Assays ›
Gene Symbol:
CDH20
Gene Name:
cadherin 20
Gene Aliases:
CDH7L3, Cdh7
Chromosome Location:
Chr.18: 61333582 - 61555773 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00906891_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031891.3 NP_114097.2 3 - 4 943 58
GenBank mRNA AF217289.1 - 2 - 3 557 58
AK290190.1 - 3 - 4 770 58
AK313809.1 - 3 - 4 822 58
BC101047.1 - 2 - 3 557 40007940 58
BC101048.1 - 2 - 3 557 40007941 58
BC101049.1 - 2 - 3 557 40007942 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800007490.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000217.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH20

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR18002466.hg.1 ENST00000262717.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002466.hg.1 ENST00000536675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002466.hg.1 ENST00000538374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002466.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002466.hg.1 TC18000217.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 TC1800007490.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800151346.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0