This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00906898_m1
See other CDH20 GE Assays ›
Gene Symbol:
CDH20
Gene Name:
cadherin 20
Gene Aliases:
CDH7L3, Cdh7
Chromosome Location:
Chr.18: 61333582 - 61555773 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00906898_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031891.3 NP_114097.2 10 - 11 2039 86
GenBank mRNA AF217289.1 - 9 - 10 1653 86
AK094366.1 - 3 - 4 261 86
AK290190.1 - 10 - 11 1866 86
AK313809.1 - 10 - 11 1918 86
BC101047.1 - 9 - 10 1653 40007940 86
BC101048.1 - 9 - 10 1653 40007941 86
BC101049.1 - 9 - 10 1653 40007942 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH20

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000262717.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000536675.2 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000538374.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 ENST00000587582.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq
Affymetrix Human Clariom D Assay TC1800007490.hg.1 NM_031891.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0