This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00907060_m1
See other NEIL3 GE Assays ›
Gene Symbol:
NEIL3
Gene Name:
nei like DNA glycosylase 3
Gene Aliases:
FGP2, FPG2, NEI3, ZGRF3, hFPG2, hNEI3
Chromosome Location:
Chr.4: 177309837 - 177362938 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00907060_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEIL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018248.2 NP_060718.2 8 - 9 1581 69
GenBank mRNA AB079071.1 - 8 - 9 1464 69
AK001720.1 - 8 - 9 1544 69
BC025954.1 - 8 - 9 1537 4853168 69
CN426741.1 - 1 - 2 223 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400009396.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000874.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NEIL3

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

base-excision repair
nucleotide-excision repair
bubble DNA binding
damaged DNA binding
double-stranded DNA binding
single-stranded DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
zinc ion binding
DNA N-glycosylase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04012010.hg.1 TC04000874.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012010.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012010.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012007.hg.1 TC04000874.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012007.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012007.hg.1 NM_018248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012006.hg.1 TC04000874.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012006.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012006.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164464.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164464.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164464.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164464.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164464.hg.1 TC0400009396.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400164463.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164463.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164463.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164463.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164463.hg.1 TC0400009396.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 ENST00000513321.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay TC0400009396.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164462.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164462.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164462.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164462.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164462.hg.1 TC0400009396.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400164461.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164461.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164461.hg.1 ENST00000264596.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400164461.hg.1 NM_018248.2 Refseq
Affymetrix Human Clariom D Assay PSR0400164461.hg.1 TC0400009396.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 172546 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0