This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00907895_m1
See other HOGA1 GE Assays ›
Gene Symbol:
HOGA1
Gene Name:
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases:
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
Chromosome Location:
Chr.10: 97584345 - 97612802 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00907895_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HOGA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138413.3 NP_612422.2 1 - 2 568 69
GenBank mRNA AK094791.1 - 1 - 2 590 69
AK289440.1 - 1 - 2 235 69
BC045550.1 - 1 - 2 552 5274464 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012489.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10002957.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HOGA1

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

glyoxylate catabolic process
4-hydroxyproline catabolic process
oxalate metabolic process
pyruvate biosynthetic process
glyoxylate metabolic process
4-hydroxy-2-oxoglutarate aldolase activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10008103.hg.1 TC10002957.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10008103.hg.1 ENST00000370646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008103.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10008102.hg.1 TC10002957.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10008102.hg.1 ENST00000370646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008102.hg.1 ENST00000370647.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008102.hg.1 NM_001134670.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10008102.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370642.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370642.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370642.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay PSR1000186114.hg.1 TC1000012489.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay PSR1000186112.hg.1 TC1000012489.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370642.4 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370642.4 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370642.4 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370646.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000370647.8 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 ENST00000465608.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_001134670.1 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq
Affymetrix Human Clariom D Assay TC1000012489.hg.1 NM_138413.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0