This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00907924_m1
See other TRAT1 GE Assays ›
Gene Symbol:
TRAT1
Gene Name:
T cell receptor associated transmembrane adaptor 1
Gene Aliases:
HSPC062, TCRIM, TRIM, pp29/30
Chromosome Location:
Chr.3: 108822698 - 108855005 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00907924_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016388.3 NP_057472.2 1 - 2 241 149
GenBank mRNA AF161547.1 - 1 - 2 156 149
AJ224878.1 - 1 - 2 140 149
AK291991.1 - 1 - 2 241 149
BC025713.1 - 1 - 2 147 5227396 149
BX415345.2 - 1 - 2 149 149

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008290.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000549.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF161547.1,AJ224878.1,AK291991.1,BC025713.1,NM_016388.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAT1

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of receptor recycling
adaptive immune response
cellular defense response
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
regulation of phosphatidylinositol 3-kinase signaling
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
positive regulation of calcium-mediated signaling
T cell receptor signaling pathway
positive regulation of T cell receptor signaling pathway
negative regulation of transport
transmembrane receptor protein tyrosine kinase adaptor activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03010083.hg.1 ENST00000295756.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010083.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010083.hg.1 ENST00000493604.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010083.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010083.hg.1 TC03000549.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03010086.hg.1 ENST00000295756.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010086.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010086.hg.1 ENST00000493604.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010086.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010086.hg.1 TC03000549.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03010085.hg.1 ENST00000295756.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010085.hg.1 ENST00000493604.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010085.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010085.hg.1 TC03000549.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010084.hg.1 ENST00000295756.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010084.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010084.hg.1 ENST00000493604.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010084.hg.1 NM_016388.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010084.hg.1 TC03000549.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay TC0300008290.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158978.hg.1 TC0300008290.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000426646.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158979.hg.1 TC0300008290.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000295756.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000478830.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000484927.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 ENST00000493604.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 NM_016388.3 Refseq
Affymetrix Human Clariom D Assay PSR0300158981.hg.1 TC0300008290.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 139804 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline