This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00908574_g1
See other FABP9 GE Assays ›
Gene Symbol:
FABP9
Gene Name:
fatty acid binding protein 9
Gene Aliases:
PERF, PERF15, T-FABP, TLBP
Chromosome Location:
Chr.8: 81458383 - 81461523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00908574_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FABP9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080526.1 NP_001073995.1 2 - 3 247 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010937.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001364.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

acrosome assembly
transport
triglyceride catabolic process
transporter activity
lipid binding

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534485 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16110594 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534446 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534494 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251698 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534448 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251712 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251741 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534502 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534498 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251737 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534503 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534462 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251707 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534481 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534459 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534465 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534470 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534438 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534476 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534486 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534488 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534468 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534454 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251706 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534461 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534477 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534466 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251720 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534463 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534499 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534471 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534440 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534489 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534484 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251704 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16110598 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534505 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534453 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16110599 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534443 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534496 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534501 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251761 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251703 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534479 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251746 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251738 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534480 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534452 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251750 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534497 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534460 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06251714 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534500 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534441 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534474 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534504 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26534467 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 468759 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348348 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1488 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1489 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 56510 - -
Agilent Human Genome CGH + SNP Microarray A_16_P38465237 - -
Agilent Human Genome CGH + SNP Microarray A_16_P32296101 - -
Agilent Human Genome CGH + SNP Microarray A_16_P32296135 - -
Agilent Human Genome CGH + SNP Microarray A_16_P32296073 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348348 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3348348 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16110598 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534477 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534479 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534466 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534494 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534486 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534488 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534454 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534468 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534476 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251737 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534459 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534485 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251761 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534490 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534489 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534467 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534461 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534463 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534502 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534505 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251750 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534457 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251738 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534499 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534504 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534496 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534484 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251746 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534471 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16110594 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534477 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534488 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16110598 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534505 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534468 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534467 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534489 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534479 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16110594 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534476 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534502 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251746 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534504 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251738 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534454 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534490 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534485 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251750 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534457 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534496 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534466 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251737 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534463 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534484 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534494 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534499 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534461 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534486 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06251761 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534459 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26534471 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18372008 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32296101 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251733 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251755 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251736 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251750 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251722 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16110588 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251735 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251760 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251699 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251745 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251659 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251728 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251716 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251664 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251737 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251748 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251702 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16110576 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251652 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251666 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251709 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251730 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251672 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251705 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16110593 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251649 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251656 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251742 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251726 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251676 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251740 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251711 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251758 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251713 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251751 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251752 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251756 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251743 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251678 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16110581 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251663 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251674 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251707 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251668 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251654 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251719 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251727 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16110579 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251731 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251650 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06251725 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18372008 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Genome U219 Array 11742668_PM_at - -
Affymetrix Human Genome U219 Array 11742668_at - -
Affymetrix Human Transcriptome Array 2.0 PSR08017595.hg.1 TC08001364.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08017595.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08017595.hg.1 NM_001080526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Clariom D Assay PSR0800174270.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800174270.hg.1 NM_001080526.1 Refseq
Affymetrix Human Clariom D Assay PSR0800174270.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800174270.hg.1 NM_001080526.1 Refseq
Affymetrix Human Clariom D Assay PSR0800174270.hg.1 TC0800010937.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010937.hg.1 ENST00000379071.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Clariom D Assay TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Clariom D Assay TC0800010937.hg.1 NM_001080526.1 Refseq
Affymetrix Human Clariom D Assay TC0800010937.hg.1 NM_001080526.1 Refseq
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A035903 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A035903 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 107648 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_2159172 NM_001080526.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2159172 NM_001080526.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2159172 NM_001080526.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2159172 NM_001080526.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2159172 NM_001080526.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2159172 NM_001080526.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2159172 NM_001080526.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1669540 NM_001080526.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2159172 NM_001080526.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0